163.89 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
UserName
Product Code. 10462012
Quantity | Price |
---|---|
1 | £ 213.0 / Pack of 50 |
Format | Standard |
---|---|
Lid | With Lid |
Culture Area | 0.38 cm2 |
Array | 8 x 12 |
Binding Type | TC-Treated |
Color | Clear |
Material | Polystyrene |
No. of Wells | 96 |
Volume (Metric) Working | 75 to 200 μL |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Total Volume (Metric) Well | 0.38 mL |
Well Shape | Conical Bottom (V-well) |
For Use With (Application) | Attachment and growth of anchorage-dependent cells |
No. per Case | 50/Cs. |
No. per Pack | 1/Pk. |
Quantity | 50/Cs. |
Diameter (Metric) Well | 1.6 cm |
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
There was an issue loading the e-mail form