183.42 GBP valid until 2025-08-31
Use promo code "24124" to get your promotional price.
Use promo code "24124" to get your promotional price.
Certifications/Compliance | ISO 9001:2000, FDA, CFR |
---|---|
Closure Material | Polyethylene |
Closure Color | Blue |
Diameter (English) Outside | 0.7 in. |
Color | Clear |
Capacity (Metric) | 15 mL |
Material | Polystyrene |
For Use With | Cell centrifugation (pelleting bacterial cells, separation by density gradients); Molecular biology (purification and precipitation of nucleic acids), Sample storage (stable 4° to 60°C); Centrifugation of precipitates |
No. per Case | 500 |
No. per Pack | 50 |
Packaging | Rack |
Sterility | Sterile |
Shape | Conical |
Length (Metric) | 120 mm |
Capacity (English) | 0.507 oz. |
Bottom Shape | Conical |
Closure Type | Dome Seal Screw Cap |
Writing Area | White Marking Spot |
Diameter (Metric) | 17 mm |
Graduated | Yes |
Product Type | Centrifuge Tube |
Max. RCF | 1800 x g |