Thermo Scientific Acros 1-Propanol, 99+%, Extra Pure
CAS: 71-23-8 | C3H8O | 60.10 g/mol
Overview
337.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 29
Additional Details:
Additional Details: CAS Number: 71-23-8 Weight: 20.00000kg Transport: UN number : 1274 Chem class : 3 Pack group : II
Quantity 25 L
Packaging Metal Drum
Disclaimers: RUO – Research Use Only
Product Code. 10469973
Quantity Price
1 £ 338.69 / 25 L
Estimated Shipment
13-10-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Chemical Identifiers
CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO
Specification
Chemical Name or Material 1-Propanol
Melting Point -127.0°C
CAS 71-23-8
CAS Min % 99.0
CAS Max % 100.0
Density 0.8000g/mL
Boiling Point 97.0°C
Quantity 25 L
Flash Point 15°C
Infrared Spectrum Authentic
Packaging Metal Drum
Physical Form Liquid
Refractive Index 1.3840 to 1.3860
Assay Percent Range 99+%
Residue after Evaporation 0.005% max.
Specific Gravity 0.8
Molecular Formula C3H8O
Linear Formula CH3CH2CH2OH
MDL Number MFCD00002941
Vapor Pressure Vapor Pressure: 25mbar at 20°C
Viscosity 2.2 mPa.s (20°C)
Beilstein 01, IV, 1413
Water 0.5% max.
Merck Index 15, 7955
Synonym 1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
Synonym 1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
Synonym 1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
Synonym 1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
Synonym 1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
Synonym 1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
Synonym 1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
Synonym 1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
Synonym 1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
Synonym 1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
SMILES CCCO
IUPAC Name propan-1-ol
Molecular Weight (g/mol) 60.10
PubChem CID 1031
ChEBI CHEBI:28831
Formula Weight 60.1
Percent Purity ≥99%
Grade Extra Pure