| Chemical Name or Material |
Hexyl alcohol
|
| Acidity |
0.02mg KOH/g max.
|
| CAS |
111-27-3
|
| Melting Point |
-52.0°C
|
| Density |
0.8136g/mL
|
| Boiling Point |
156.0°C to 157.0°C
|
| Flash Point |
59°C
|
| Infrared Spectrum |
Authentic
|
| Assay Percent Range |
97.5% min. (GC)
|
| Packaging |
Glass bottle
|
| Molecular Formula |
C6H14O
|
| Linear Formula |
CH3(CH2)5OH
|
| Refractive Index |
1.4170 to 1.4190
|
| MDL Number |
MFCD00002982
|
| Quantity |
1 L
|
| Beilstein |
01, 407
|
| Merck Index |
15, 4734
|
| Specific Gravity |
0.8136
|
| Synonym |
1-hexanol,hexyl alcohol,hexanol,n-hexanol,n-hexyl alcohol,1-hexyl alcohol,1-hydroxyhexane,amylcarbinol,caproyl alcohol,pentylcarbinol
|
| Synonym |
1-hexanol,hexyl alcohol,hexanol,n-hexanol,n-hexyl alcohol,1-hexyl alcohol,1-hydroxyhexane,amylcarbinol,caproyl alcohol,pentylcarbinol
|
| Synonym |
1-hexanol,hexyl alcohol,hexanol,n-hexanol,n-hexyl alcohol,1-hexyl alcohol,1-hydroxyhexane,amylcarbinol,caproyl alcohol,pentylcarbinol
|
| Synonym |
1-hexanol,hexyl alcohol,hexanol,n-hexanol,n-hexyl alcohol,1-hexyl alcohol,1-hydroxyhexane,amylcarbinol,caproyl alcohol,pentylcarbinol
|
| Synonym |
1-hexanol,hexyl alcohol,hexanol,n-hexanol,n-hexyl alcohol,1-hexyl alcohol,1-hydroxyhexane,amylcarbinol,caproyl alcohol,pentylcarbinol
|
| Synonym |
1-hexanol,hexyl alcohol,hexanol,n-hexanol,n-hexyl alcohol,1-hexyl alcohol,1-hydroxyhexane,amylcarbinol,caproyl alcohol,pentylcarbinol
|
| Synonym |
1-hexanol,hexyl alcohol,hexanol,n-hexanol,n-hexyl alcohol,1-hexyl alcohol,1-hydroxyhexane,amylcarbinol,caproyl alcohol,pentylcarbinol
|
| Synonym |
1-hexanol,hexyl alcohol,hexanol,n-hexanol,n-hexyl alcohol,1-hexyl alcohol,1-hydroxyhexane,amylcarbinol,caproyl alcohol,pentylcarbinol
|
| Synonym |
1-hexanol,hexyl alcohol,hexanol,n-hexanol,n-hexyl alcohol,1-hexyl alcohol,1-hydroxyhexane,amylcarbinol,caproyl alcohol,pentylcarbinol
|
| Synonym |
1-hexanol,hexyl alcohol,hexanol,n-hexanol,n-hexyl alcohol,1-hexyl alcohol,1-hydroxyhexane,amylcarbinol,caproyl alcohol,pentylcarbinol
|
| Solubility Information |
Solubility in water: 6g/L (25°C). Other solubilities: miscible with alcohol and ether
|
| InChI Key |
ZSIAUFGUXNUGDI-UHFFFAOYSA-N
|
| SMILES |
CCCCCCO
|
| IUPAC Name |
hexan-1-ol
|
| Molecular Weight (g/mol) |
102.18
|
| PubChem CID |
8103
|
| ChEBI |
CHEBI:87393
|
| Viscosity |
5.3 mPa.s (20°C)
|
| Formula Weight |
102.18
|
| Percent Purity |
98%
|
| Grade |
Pure
|
| Physical Form |
Liquid
|