Kautex™ Polyethylene Wide Neck Jars
Manufactured from HDPE for strength and durability. Kautex™ Polyethylene Wide Neck Jars provide easy to fill openings to minimize waste.
Overview
Code: NEW
Additional Details:
Additional Details: Weight: 0.01800kg
Product Code. 10490781
Quantity Price
1 £ 1.75 / Each
Estimated Shipment
13-08-2025
Add to basket
  • Description and Specification
  • Documentation
  • Accessories and Spare Parts
Description and Specification
Specification
Capacity (Metric) 250 mL
Color Natural
Diameter (Metric) Inner Neck 41 mm
For Use With (Application) Dispensing, Packaging, Storage
Material HDPE
No. per Case 298/Cs.
Weight (Metric) 0.02 kg
Clarity Natural
Closure Included No
Mouth Wide
Shape Round
Height (Metric) 98 mm
Product Type Storage Container
Diameter (Metric) 65 mm
  • Round, sturdy, easy to clean and leakproof
  • Particularly suitable for storage, transport or disposal of powdery or granulated materials
  • Closures are not included
Applications:
  • packaging
  • dispensing
  • sending samples
  • mailing
  • transport
  • storage
Documentation Accessories and Spare Parts