684.00 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Use promo code "24155" to get your promotional price.
Orbit | 4.5 mm |
---|---|
Speed Range | 0 to 3000 rpm |
Electrical Requirements | 100/240 V, 50/60 Hz, 20 W |
Voltage | 100/240 V |
Wattage | 20 W |
Amperage | 800 mA |
Product Type | Vortex Mixer |
Certifications/Compliance | DIN EN 60529 |
Frequency | 50/60 Hz |
Dimensions (L x W x H) | 205 x 148 x 63 mm |
Height (Metric) | 63 mm |
Width (Metric) | 148 mm |
Weight (Metric) | 2.9 kg |
Model | MS 3 Basic |
Display | Scale |
IP Rating | IP 21 |
Motor Type | Orbital |
Operating Modes | Continuous |