787.67 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Material | Polystyrene |
---|---|
Volume (Metric) Well | 400 μL |
Volume (Metric) Working | 50 to 200 μL |
Type | Optical Polymer Base |
Quantity | Case of 20 |
No. per Pack | 5 |
No. per Case | 20 |
Color | Black |
Surface Treatment | Poly-D-Lysine |
Sterility | Aseptically Manufactured |
No. of Wells | 96 |
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Array | 8 x 12 |
Product Line | Nunc™ |
Packaging | 5/Pack, 20/Case |
Barcode | Custom Barcoding Available |
Well Shape | Flat |