Use promo code "25339" to get your promotional price.
Thermo Scientific Acros Diethyl ether, 99+%, extra pure, stabilized with BHT
Use promo code "25339" to get your promotional price.
Quantity | 1 L |
---|---|
Packaging | Glass Bottle |
Chemical Identifiers
CAS | 60-29-7 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | MFCD00011646 |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
Synonym | diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether |
PubChem CID | 3283 |
ChEBI | CHEBI:35702 |
IUPAC Name | ethoxyethane |
SMILES | CCOCC |
Specification
Chemical Name or Material | Diethyl ether |
---|---|
Melting Point | -116.0°C |
Name Note | Stabilized with BHT |
CAS | 60-29-7 |
Acidity | 2mg/100g max. |
Density | 0.7140g/mL |
Boiling Point | 34.6°C |
Quantity | 1 L |
Flash Point | −45°C |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.3510 to 1.3530 |
Assay Percent Range | 99+% |
Residue after Evaporation | 0.005% max. |
Specific Gravity | 0.714 |
Molecular Formula | C4H10O |
Linear Formula | (CH3CH2)2O |
MDL Number | MFCD00011646 |
Viscosity | 0.2448 mPa.s (20°C) |
Water | 0.1% max. (Coulometric) |
Synonym | diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether |
Synonym | diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether |
Synonym | diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether |
Synonym | diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether |
Synonym | diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether |
Synonym | diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether |
Synonym | diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether |
Synonym | diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether |
Synonym | diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether |
Synonym | diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether |
Solubility Information | Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid, miscible with lower aliphatic alcohols, benzene,, chloroform, petroleum ether and many oils |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
SMILES | CCOCC |
IUPAC Name | ethoxyethane |
Molecular Weight (g/mol) | 74.12 |
PubChem CID | 3283 |
ChEBI | CHEBI:35702 |
Formula Weight | 74.12 |
Percent Purity | ≥99% |
Grade | Extra Pure |
Technical documents
[Diethyl ether]
[Danger]
[{code=FLAM. LIQ. 1, sequence=1, description=Flammable liquid Category 1}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=STOT SE 3, sequence=3, description=Specific target organ toxicity Category 3}]
[{code=H224, sequence=1, description=Extremely flammable liquid and vapour.}, {code=H302, sequence=2, description=Harmful if swallowed.}, {code=H336, sequence=3, description=May cause drowsiness or dizziness.}, {code=EUH019, sequence=91, description=May form explosive peroxides.}, {code=EUH066, sequence=92, description=Repeated exposure may cause skin dryness or cracking.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P233, sequence=52, description=Keep container tightly closed.}, {code=P240, sequence=53, description=Ground/bond container and receiving equipment.}, {code=P243, sequence=54, description=Take precautionary measures against static discharge.}, {code=P261, sequence=55, description=Avoid breathing dust/fume/gas/mist/vapours/spray.}, {code=P301+P312, sequence=56, description=IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.}, {code=P403+P235, sequence=57, description=Store in a well-ventilated place. Keep cool.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- Diethyl ether
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
Hazard Statement
- H224- Extremely flammable liquid and vapour.
- H302- Harmful if swallowed.
- H336- May cause drowsiness or dizziness.
- EUH019- May form explosive peroxides.
- EUH066- Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233- Keep container tightly closed.
- P240- Ground/bond container and receiving equipment.
- P243- Take precautionary measures against static discharge.
- P261- Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312- IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P403+P235- Store in a well-ventilated place. Keep cool.