Macherey-Nagel™ pH Neutral Litmus Paper Strips
Additional Details:
Weight:
0.00900kg
Specification
Color Indicator | Red to Violet to Blue |
---|---|
pH Range | 8 to 5 |
Format | Strip |
For Use With (Application) | Determination of acidic and alkaline reaction |
Quantity | 100 Pack |
Type | pH Paper |