797.10 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Use for analyzing difficult-to-resolve mixtures of halogenated compounds including pesticides.
Column Format | Analytical Column |
---|---|
Diameter (Metric) | 3 mm |
For Use With | HPLC |
Length (Metric) | 100 mm |
Max. Pressure | 18130 psi (1250 bar) |
Particle Size | 1.9 μm |
pH | 2 to 8 |
Pore Size | 175 Å |
Surface Area | 220 m²/g |
Temperature | 60°C |
USP Type | L43 |
Stationary Phase | PFP (Pentafluorophenylpropyl) |
Product Line | Hypersil GOLD |
Packing Material | Spherical, Fully Porous, Ultrapure Silica |
Carbon Load | 8% |
Column Type | Reversed Phase |
Endcapped | Yes |
Phase | Reversed Phase |