Thermo Scientific™ Pierce™ Bovine Serum Albumin Standard Pre-Diluted Set
High-quality, calibrated BSA solutions (2mg/mL) to serially dilute to create protein assay standard curves to accurately measure protein concentration.
Overview
122.00 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Code: PN
Additional Details:
Additional Details: Weight: 1.81440kg
Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Product Code. 10531674
Quantity Price
1 £ 183.0 / 24.50mL
In stock, ready to ship: 15
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Special Offers
Description and Specification
Specification
Content And Storage BSA in 0.9% sodium chloride with sodium azide; one each of the following μg/mL concentrations: 125, 250, 500, 750, 1000, 1500, 2000
Store at 2–8°C.
Description Pierce Bovine Serum Albumin Standard Pre-Diluted Set
Assay Protein Standards
Product Line Pierce™
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
For Use With (Application) Internal Standard
Quantity 7 x 3.5 mL
These bovine serum albumin (BSA) solutions are protein concentration reference standards for use in BCA, Bradford and other protein assay protocols. BSA is the universally accepted reference protein for total protein quantitation. The albumin standard is precisely formulated at 2mg/mL in an ultrapure 0.9% sodium chloride (saline) solution. The concentration of the stock solution is calibrated by direct comparison to purified BSA (Fraction V) from the National Institute of Standards and Technology (NIST™). The product is available in three convenient package formats: 1mL glass ampules, 50mL polypropylene bottles, and complete sets containing seven ready-to-use dilutions.
Highlights:
  • Universal – recognized as the industry standard for protein quantitation in colorimetric protein assays
  • Pure and stable – supplied in ultrapure 0.9% saline solution with 0.05% sodium azide; room temperature stable
  • Accurate and consistent – precisely formulated at 2.00 +/-0.03mg/mL compared to a NIST reference
  • Convenient – choose 1mL ampules, a 50mL bottle, or a complete set containing seven ready-to-use dilutions
Includes:
2mg/mL BSA solution (Part No. 23209, 23210) or 7-solution set of different concentrations (Part No. 23208)
Recommended for:
Protein assay quantitation standard; Protein recovery control for desalting and other column procedures; General calibration of spectrophotometer UV-lamp (A280)