89.49 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Use promo code "25721" to get your promotional price.
Barcode | No Barcode |
---|---|
Format | Standard |
Array | 8 x 12 |
Autoclavable | Autoclavable |
Color | Clear |
Product Type | Deep-well Plate |
Material | Polypropylene |
Volume (Metric) Well | 1.1 mL |
No. of Wells | 96 |
Sterility | Non-sterile |
Surface Treatment | Non-treated |
Well Shape | Round |
For Use With (Application) | Flexible mat covers or adhesive sealing films to reduce sample evaporation and contamination |
No. per Pack | 24 |
Profile | U-bottom |
Quantity | 24 Pk. |