Chemical Name or Material |
Methyl sulfoxide
|
Melting Point |
18.4°C
|
CAS |
67-68-5
|
Color |
Brown to White
|
Decomposition Information |
>190°C
|
Density |
1.1000g/mL
|
Boiling Point |
189.0°C
|
Quantity |
2.5 L
|
Flash Point |
87°C
|
Infrared Spectrum |
Authentic
|
Packaging |
AcroSeal™ Glass Bottle
|
Physical Form |
Powder, Needles or Crystals
|
Refractive Index |
1.4770 to 1.4800
|
Assay Percent Range |
99.7% min. (GC)
|
Specific Gravity |
1.1
|
Molecular Formula |
C2H6OS
|
Linear Formula |
(CH3)2SO
|
MDL Number |
MFCD00002089
|
Vapor Pressure |
0.55mbar at 20°C
|
Viscosity |
1.987 mPa.s (25°C)
|
Beilstein |
01, IV, 1277
|
Water |
0.005% Max.
|
Fieser |
01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
|
Merck Index |
15, 3285
|
Synonym |
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
|
Synonym |
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
|
Synonym |
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
|
Synonym |
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
|
Synonym |
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
|
Synonym |
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
|
Synonym |
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
|
Synonym |
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
|
Synonym |
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
|
Synonym |
dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide
|
Solubility Information |
Solubility in water: soluble. Other solubilities: soluble in alcohol,benzene,acetone,chloroform,,diethyl ether and aromatic hydrocarbons
|
InChI Key |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
|
SMILES |
CS(C)=O
|
IUPAC Name |
methylsulfinylmethane
|
Molecular Weight (g/mol) |
78.13
|
PubChem CID |
679
|
ChEBI |
CHEBI:28262
|
Formula Weight |
78.13
|
Percent Purity |
99.7+%
|
Grade |
Extra Dry over Molecular Sieve
|