Thermo Scientific™ Salmonella Shigella Agar (SS Agar) / XLD Agar Biplate
Isolate and identify Salmonella and Shigella spp. from clinical or food samples with highly selective media biplates which aid recovery from mixed flora specimens.
Overview
13.60 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: Weight: 0.50000kg
Product Code. 10557163
Quantity Price
1 £ 23.85 / Pack of 10
Estimated Shipment
05-06-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Specification
Content And Storage 2° to 12°C, in the dark
Description Salmonella Shigella Agar (SS Agar) / XLD Agar
Format 90mm biplate
Product Type Agar
Fill Weight 17.5g ± 5%
Packaging Type Wrapped in single cellulose-based film wrap
Quantity 10/Pk.
Salmonella and Shigella are major causes of bacterial enteric illness with transmission occuring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water. Salmonella Shigella Agar (SS Agar) / XLD Agar is ideal for screening samples containing mixed flora and suspected of harbouring enteric pathogens, e.g., medical specimens or food products.
  • Sensitivity and selectivity improves recovery of salmonellae and shigellae
  • Ready-to-use convenience of prepared media
  • XLD Agar is recommended in the ISO 6579 testing of Salmonella
Widely recognised in international standards, XLD Agar, relies on xylose fermentation, lysine decarboxylation and production of hydrogen sulphide for the primary differentiation of Shigella and Salmonella spp. from non-pathogenic bacteria. The sensitivity and selectivity of XLD Agar exceeds that of the traditional plating media, e.g., Eosin Methylene Blue and Bismuth Sulphite agars, which tend to suppress the growth of shigellae.
Our formulation of Salmonella Shigella Agar (SS Agar) has been modified from the original for improved performance. Although widely used, traditional SS Agar has been criticized because of excessive inhibition of Shigella species. Alterations to the bile salt mixture, peptone and pH value considerably improve the growth of shigellae without too much increased growth of commensal organisms. Salmonella colonies are also larger with improved blackening at the centre to aid identification.
XLD Agar is recommended in the ISO 6579 testing of Salmonella and is tested according to ISO 11133-2.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.