Code:
04
Hanna Instruments pH 4.01 Calibration Buffer Solution Sachets, Hanna Instruments™
Premium quality pH 4.01 calibration buffer that is NIST traceable and supplied with a Certificate of Analysis that is ideal or users that do not calibrate frequently.
Additional Details:
Weight:
0.50000kg
Quantity | 20 mL |
---|
Specification
Color | Undesignated |
---|---|
pH | 4.01 |
Physical Form | Liquid |
Quantity | 20 mL |
Formulation | Aqueous Buffer Solution |
Odor | Odorless |
Packaging | 25Pack |
Traceability to NIST | Yes |