Use promo code "24156" to get your promotional price.
Fisher Chemical Tetrahydrofuran, for HPLC, Unstabilised, Fisher Chemical™
Use promo code "24156" to get your promotional price.
Quantity | 2.5 L |
---|---|
Packaging | Plastic-coated Glass Bottle |
Chemical Identifiers
CAS | 109-99-9 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.11 |
MDL Number | MFCD00005356 |
InChI Key | WYURNTSHIVDZCO-UHFFFAOYSA-N |
Synonym | tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano |
PubChem CID | 8028 |
ChEBI | CHEBI:26911 |
IUPAC Name | oxolane |
SMILES | C1CCOC1 |
Specification
pH | 7 to 8 |
---|---|
Melting Point | -108.4°C |
CAS | 109-99-9 |
Color | Colorless |
Boiling Point | 66°C |
Quantity | 2.5 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Molecular Formula | C4H8O |
MDL Number | MFCD00005356 |
Vapor Pressure | 200mbar at 20°C |
Viscosity | 0.55cp at 20°C |
Synonym | tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano |
Synonym | tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano |
Synonym | tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano |
Synonym | tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano |
Synonym | tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano |
Synonym | tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano |
Synonym | tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano |
Synonym | tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano |
Synonym | tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano |
Synonym | tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano |
InChI Key | WYURNTSHIVDZCO-UHFFFAOYSA-N |
SMILES | C1CCOC1 |
IUPAC Name | oxolane |
Molecular Weight (g/mol) | 72.11 |
PubChem CID | 8028 |
ChEBI | CHEBI:26911 |
Formula Weight | 72.11g/mol |
Technical documents
[Tetrahydrofuran]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=EYE DAM. 2, sequence=3, description=Serious eye damage/eye irritation Category 2}, {code=STOT SE 3, sequence=4, description=Specific target organ toxicity Category 3}, {code=CARC. 2, sequence=5, description=Carcinogenicity Category 2}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H302, sequence=2, description=Harmful if swallowed.}, {code=H319, sequence=3, description=Causes serious eye irritation.}, {code=H335, sequence=4, description=May cause respiratory irritation.}, {code=H336, sequence=5, description=May cause drowsiness or dizziness.}, {code=H351, sequence=6, description=Suspected of causing cancer.}, {code=EUH019, sequence=91, description=May form explosive peroxides.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P303+P361+P353, sequence=52, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P312, sequence=54, description=Call a POISON CENTER/doctor/if you feel unwell.}, {code=P304+P340, sequence=55, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P280, sequence=56, description=Wear protective gloves/protective clothing/eye protection/face protection.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Tetrahydrofuran}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}, {code=GHS08, sequence=3, description=HEALTH HAZARD}]
Product Identifier
- Tetrahydrofuran
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H302- Harmful if swallowed.
- H319- Causes serious eye irritation.
- H335- May cause respiratory irritation.
- H336- May cause drowsiness or dizziness.
- H351- Suspected of causing cancer.
- EUH019- May form explosive peroxides.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312- Call a POISON CENTER/doctor/if you feel unwell.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental Information
- MIXTURE LIST- Contains : Tetrahydrofuran