56.78 GBP valid until 2025-06-30
Use promo code "24108" to get your promotional price.
Use promo code "24108" to get your promotional price.
Concentration | 6X |
---|---|
Gel Type | Agarose |
Label or Dye | Xylene Cyanol FF, Bromophenol Blue, Orange G |
Product Type | DNA Ladder |
Ready to Load | Yes |
Size Range | 100 to 5000 bp |
Gel Compatibility | Agarose Gels |
Green Features | Sustainable packaging |
Product Line | GeneRuler |
Quantity | 50 μg |