Thermo Scientific™ NotI, HC (50 U/μL)
Thermo Scientific™ Fermentas NotI is available as a FastDigest enzyme which is used for rapid DNA digestion.
Overview
138.00 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Code: 88
Additional Details:
Additional Details: Weight: 1.51950kg
Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Product Code. 10591031
Quantity Price
1 £ 207.0 / 1500 units
Estimated Shipment
30-07-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Concentration 50 U/μL
Methylation Sensitivity CpG Methylation-Sensitive,Not Dam Methylation-Sensitive,Not Dcm Methylation-Sensitive
Methylation Sensitivity CpG Methylation-Sensitive,Not Dam Methylation-Sensitive,Not Dcm Methylation-Sensitive
Methylation Sensitivity CpG Methylation-Sensitive,Not Dam Methylation-Sensitive,Not Dcm Methylation-Sensitive
Enzyme Not I
Compatible Buffer 10X Buffer O
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 1,500 units
Product Type Restriction Enzyme
Research Category Traditional Cloning
5'..G C▵G G C C G C...3'
3'..C G C C G G▵C G...5'
Conditions for 100% Activity:
  • 1X Buffer O: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
  • Incubate at 37°C
Storage Buffer:
  • NotI is supplied in: 20mM Tris-HCl (pH 7.8 at 25°C), 100mM NaCl, 0.1mM EDTA, 1mM DTT, 0.02% Triton X-100, 0.2mg/mL BSA and 50% (v/v) glycerol
Ligation and Recleavage:
  • After 50-fold overdigestion with NotI, more than 95% of DNA fragments can be ligated and recut
Methylation Effects:
  • Dam: never overlaps —no effect
  • Dcm: never overlaps —no effect
  • CpG: completely overlaps —blocked
  • EcoKI: never overlaps —no effect
  • EcoBI: never overlaps —no effect
Digestion of Agarose-embedded DNA:
  • Minimum 5units of the enzyme are required for complete digestion of 1μg of agarose-embedded Adenovirus-2 DNA in 16 hours
Compatible Ends:
  • Bsp120I, CfrI, Eco52I
Note:
Assayed using pTZ19RJL2 plasmid DNA digested with BseLI. Supercoiled plasmids may require up to 5-fold more NotI for complete digestion than linear DNAs.