121.64 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Use promo code "24155" to get your promotional price.
UserName
Product Code. 10595025
Quantity | Price |
---|---|
1 | £ 145.0 / Pack of 100 |
Color | Clear |
---|---|
Material | Polyethylene |
Sterility | Non-sterile |
No. of Wells | 96 |
Quantity | 100 films |
Content And Storage |
Contains: 100 films Storage: Room temperature |
For Use With (Equipment) | GeneAmp 9700, SimpliAmp™ Thermal Cycler, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ProFlex™ PCR System |
Shipping Condition | Room Temperature |
Product Type | Film, Seal, or Applicator |
Product Line | MicroAmp |
Adhesive | Adhesive Film |
Temperature Range (Metric) | -20°C to +120°C |
Plate Compatibility | 96/384-well Plate |
Available as clear (economical and sticky) or optical (ready for real-time PCR and nontacky)
Reduce Leaks, Evaporation, and Well-to-Well Contamination
See user's manual or package insert for limited label license and trademark information.
Automated Genomic DNA Purification, Automated Nucleic Acid Purification, DNA & RNA Purification & Analysis, PCR, PCR & Real-Time PCR
There was an issue loading the e-mail form