JT Baker Acetonitrile, Ultra Gradient Grade, BAKER ANALYZED HPLC, J.T. Baker™
Quantity | 5 L |
---|---|
Packaging | EcoTainer Metal Safety Can |
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Weight (g/mol) | 41.053 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Specification
Chemical Name or Material | Acetonitrile |
---|---|
Melting Point | -46°C |
Absorbance | Max. 0.05 at 200nm,Max. 0.01 at 220nm,Max. 0.01 at 254-400nm |
CAS | 75-05-8 |
Absorbance | Max. 0.05 at 200nm,Max. 0.01 at 220nm,Max. 0.01 at 254-400nm |
Absorbance | Max. 0.05 at 200nm,Max. 0.01 at 220nm,Max. 0.01 at 254-400nm |
Color | Colorless |
Appearance | Passes test |
Boiling Point | 81°C |
Quantity | 5 L |
Explosion Limit | 4.4 - 16 Vol % |
Flash Point | 5°C |
Identification | Passes test |
Ignition Point | 525°C |
Odor | Aromatic |
Packaging | EcoTainer Metal Safety Can |
Physical Form | Liquid |
Purity Grade Notes | Ultra Gradient HPLC Grade |
Refractive Index | 1.344 n20/d |
Residue after Evaporation | Max. 1 ppm |
Molecular Formula | C2H3N |
Titratable Acid | Max. 0.0008meq/g |
Linear Formula | CH3CN |
Titratable Base | Max. 0.0006meq/g |
Viscosity | 0.39 mPas at 20°C |
Water | Max. 0.01% |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Synonym | methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitrile,cyanure de methyl,methylkyanid,mecn |
Solubility Information | Fully miscible in water |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
SMILES | CC#N |
IUPAC Name | acetonitrile |
Molecular Weight (g/mol) | 41.053 |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
Formula Weight | 41.05g/mol |
Percent Purity | ≥99.9% |
Grade | BAKER ANALYZED™ HPLC |
[Acetonitrile]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=EYE DAM. 2, sequence=3, description=Serious eye damage/eye irritation Category 2}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H302, sequence=2, description=Harmful if swallowed.}, {code=H312, sequence=3, description=Harmful in contact with skin.}, {code=H332, sequence=4, description=Harmful if inhaled.}, {code=H319, sequence=5, description=Causes serious eye irritation.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P303+P361+P353, sequence=52, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=53, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H302- Harmful if swallowed.
- H312- Harmful in contact with skin.
- H332- Harmful if inhaled.
- H319- Causes serious eye irritation.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.