Use promo code "24127" to get your promotional price.
Fisher Chemical Acetonitrile, for HPLC-MS
Use promo code "24127" to get your promotional price.
Quantity | 2.5 L |
---|
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
MDL Number | 1878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Specification
Packaging | Amber glass bottle |
---|---|
CAS | 75-05-8 |
Quantity | 2.5 L |
Vapor Pressure | 97mbar at 20°C |
Viscosity | 0.36cp at 20°C |
Vapor Density | 1.42 |
Molecular Formula | C2H3N |
Physical Form | Liquid |
MDL Number | 1878 |
Color | Colorless |
Melting Point | -46°C |
Boiling Point | 81°C |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
SMILES | CC#N |
IUPAC Name | acetonitrile |
Molecular Weight (g/mol) | 41.053 |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
Formula Weight | 41.05g/mol |
Technical documents
[Acetonitrile]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=EYE DAM. 2, sequence=3, description=Serious eye damage/eye irritation Category 2}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H319, sequence=2, description=Causes serious eye irritation.}, {code=H332, sequence=3, description=Harmful if inhaled.}, {code=H302, sequence=4, description=Harmful if swallowed.}, {code=H312, sequence=5, description=Harmful in contact with skin.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=52, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P301+P312, sequence=53, description=IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.}, {code=P304+P340, sequence=54, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P210, sequence=56, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P240, sequence=57, description=Ground/bond container and receiving equipment.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Acetonitrile}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H319- Causes serious eye irritation.
- H332- Harmful if inhaled.
- H302- Harmful if swallowed.
- H312- Harmful in contact with skin.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.
- P301+P312- IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240- Ground/bond container and receiving equipment.
Supplemental Information
- MIXTURE LIST- Contains : Acetonitrile
-
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.
Buy Now