Use promo code "25339" to get your promotional price.
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, 99.5%, ACS reagent
Use promo code "25339" to get your promotional price.
Quantity | 1 kg |
---|---|
Packaging | Plastic bottle |
Chemical Identifiers
CAS | 60-00-4 |
---|---|
Molecular Formula | C10H16N2O8 |
Molecular Weight (g/mol) | 292.24 |
MDL Number | MFCD00003541 |
InChI Key | KCXVZYZYPLLWCC-UHFFFAOYSA-N |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
PubChem CID | 6049 |
ChEBI | CHEBI:42191 |
IUPAC Name | 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid |
SMILES | OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Specification
Chemical Name or Material | Ethylenediaminetetraacetic acid |
---|---|
CAS | 60-00-4 |
Decomposition Information | >150°C |
Melting Point | 220.0°C |
Color | White |
Flash Point | >100°C |
Ignition Residue | 0.2% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 99.50% |
Packaging | Plastic bottle |
Molecular Formula | C10H16N2O8 |
Linear Formula | (HO2CCH2)2NCH2CH2N(CH2CO2H)2 |
MDL Number | MFCD00003541 |
Quantity | 1 kg |
Fieser | 01,373 |
Merck Index | 15, 3565 |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Solubility Information | 0.005% max. Insoluble Matter (in dilute NH4OH) |
InChI Key | KCXVZYZYPLLWCC-UHFFFAOYSA-N |
SMILES | OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
IUPAC Name | 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid |
Molecular Weight (g/mol) | 292.24 |
PubChem CID | 6049 |
ChEBI | CHEBI:42191 |
Formula Weight | 292.23 |
Vapor Pressure | 0.013hPa at 20°C |
Percent Purity | 99.5% |
Grade | ACS Reagent |
Physical Form | Fine Crystalline Powder |
Technical documents
[Ethylenediaminetetraacetic acid]
[Warning]
[{code=ACUTE TOX. 4, sequence=1, description=Acute toxicity Category 4}, {code=EYE DAM. 2, sequence=2, description=Serious eye damage/eye irritation Category 2}, {code=STOT RE 2, sequence=3, description=Specific target organ toxicity after repeated exposure Category 2}]
[{code=H319, sequence=1, description=Causes serious eye irritation.}, {code=H332, sequence=2, description=Harmful if inhaled.}, {code=H373, sequence=3, description=May cause damage to organs.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P304+P340, sequence=52, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=53, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P312, sequence=54, description=Call a POISON CENTER/doctor/if you feel unwell.}, {code=P337+P313, sequence=55, description=If eye irritation persists: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Ethylenediaminetetraacetic acid}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}, {code=GHS08, sequence=2, description=HEALTH HAZARD}]
Product Identifier
- Ethylenediaminetetraacetic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H319- Causes serious eye irritation.
- H332- Harmful if inhaled.
- H373- May cause damage to organs.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312- Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313- If eye irritation persists: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contains : Ethylenediaminetetraacetic acid