605.15 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Capacity (English) | 2.36 oz. |
---|---|
Closure Color | Orange |
Closure Material | HDPE |
Capacity (Metric) | 70 mL |
Material | Polystyrene |
For Use With (Application) | For optimal cell attachment |
Includes | Lot numbers for ease in traceability |
Neck Style | Canted |
Shape | Rectangular |
Surface Coating | Tissue Culture Treated |
Graduated | Yes |
Sterility | Sterile |
Closure Type | Plug Seal |
Culture Area | 25 cm² |