Use promo code "25339" to get your promotional price.
Fisher Chemical Chloroform, 99+%, Extra Pure, Stabilised with Amylene, SLR, Fisher Chemical™
Use promo code "25339" to get your promotional price.
Quantity | 5 L |
---|---|
Packaging | HDPE Bottle |
Chemical Identifiers
CAS | 67-66-3 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 119.37 |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
Synonym | |
PubChem CID | |
ChEBI | |
SMILES | ClC(Cl)Cl |
Specification
Melting Point | -63°C |
---|---|
CAS | 67-66-3 |
Color | Colorless |
Boiling Point | 61°C |
Quantity | 5 L |
Packaging | HDPE Bottle |
Physical Form | Liquid |
Molecular Formula | CHCl3 |
MDL Number | 826 |
Vapor Pressure | 213mbar at 20°C |
Viscosity | 0.56 mPaS at 20°C |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
SMILES | ClC(Cl)Cl |
IUPAC Name | trichloromethane |
Molecular Weight (g/mol) | 119.37 |
Formula Weight | 119.38g/mol |
Technical documents
[Chloroform, stabilized with amylene]
[Danger]
[{code=ACUTE TOX. 3, sequence=1, description=Acute toxicity Category 3}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=CARC. 2, sequence=3, description=Carcinogenicity Category 2}, {code=EYE DAM. 2, sequence=4, description=Serious eye damage/eye irritation Category 2}, {code=REPR. 2, sequence=5, description=Reproductive toxicity Category 2}, {code=SKIN IRRIT. 2, sequence=6, description=Skin corrosion/irritation Category 2}, {code=STOT RE 1, sequence=7, description=Specific target organ toxicity Category 1}, {code=STOT SE 3, sequence=8, description=Specific target organ toxicity Category 3}]
[{code=H372, sequence=1, description=Causes damage to organs.}, {code=H315, sequence=2, description=Causes skin irritation.}, {code=H319, sequence=3, description=Causes serious eye irritation.}, {code=H351, sequence=4, description=Suspected of causing cancer.}, {code=H361d, sequence=5, description=Suspected of damaging the unborn child.}, {code=H331, sequence=6, description=Toxic if inhaled.}, {code=H336, sequence=7, description=May cause drowsiness or dizziness.}, {code=H302, sequence=8, description=Harmful if swallowed.}]
[{code=P260, sequence=50, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=52, description=IF ON SKIN: Wash with plenty of water/.}, {code=P304+P340, sequence=53, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P311, sequence=55, description=Call a POISON CENTER/doctor/ .}]
[{code=GHS06, sequence=1, description=SKULL & CROSSBONES}, {code=GHS08, sequence=2, description=HEALTH HAZARD}]
Product Identifier
- Chloroform, stabilized with amylene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H372- Causes damage to organs.
- H315- Causes skin irritation.
- H319- Causes serious eye irritation.
- H351- Suspected of causing cancer.
- H361d- Suspected of damaging the unborn child.
- H331- Toxic if inhaled.
- H336- May cause drowsiness or dizziness.
- H302- Harmful if swallowed.
Precautionary Statement
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311- Call a POISON CENTER/doctor/ .
-
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular products
Buy Now