Use promo code "25339" to get your promotional price.
Thermo Scientific Acros Triethylamine, 99.7%, extra pure, Thermo Scientific Chemicals
Use promo code "25339" to get your promotional price.
Quantity | 2.5 kg |
---|
Chemical Identifiers
CAS | 121-44-8 |
---|---|
MDL Number | MFCD00009051 |
InChI Key | ZMANZCXQSJIPKH-UHFFFAOYSA-N |
Synonym | triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane |
PubChem CID | 8471 |
ChEBI | CHEBI:35026 |
IUPAC Name | N,N-diethylethanamine |
SMILES | CCN(CC)CC |
Specification
Chemical Name or Material | Triethylamine |
---|---|
Name Note | 99.7% |
CAS | 121-44-8 |
Melting Point | -115.0°C |
Density | 0.7280g/mL |
Boiling Point | 90.0°C |
Flash Point | −11°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99.6% min. (GC) |
Packaging | Plastic Bottle |
Molecular Formula | C6H15N |
Linear Formula | (C2H5)3N |
Refractive Index | 1.3990 to 1.4010 |
MDL Number | MFCD00009051 |
Quantity | 2.5 kg |
Beilstein | 04, 99 |
Fieser | 01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336 |
Merck Index | 15, 9831 |
Specific Gravity | 0.728 |
Synonym | triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane |
Synonym | triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane |
Synonym | triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane |
Synonym | triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane |
Synonym | triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane |
Synonym | triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane |
Synonym | triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane |
Synonym | triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane |
Synonym | triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane |
Synonym | triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane |
Solubility Information | Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride |
InChI Key | ZMANZCXQSJIPKH-UHFFFAOYSA-N |
SMILES | CCN(CC)CC |
IUPAC Name | N,N-diethylethanamine |
Molecular Weight (g/mol) | 101.19 |
PubChem CID | 8471 |
ChEBI | CHEBI:35026 |
Viscosity | 0.36 mPa.s (20°C) |
Formula Weight | 101.19 |
Vapor Pressure | 69mbar at 20°C |
Percent Purity | 99.7% |
Grade | Extra Pure |
Physical Form | Liquid |
Technical documents
[Triethylamine]
[Danger]
[{code=ACUTE TOX. 3, sequence=1, description=Acute toxicity Category 3}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=EYE DAM. 1, sequence=3, description=Serious eye damage/eye irritation Category 1}, {code=FLAM. LIQ. 2, sequence=4, description=Flammable liquid Category 2}, {code=SKIN CORR. 1A, sequence=5, description=Skin corrosion/irritation Category 1 A}, {code=STOT SE 3, sequence=6, description=Specific target organ toxicity Category 3}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H302, sequence=2, description=Harmful if swallowed.}, {code=H311, sequence=3, description=Toxic in contact with skin.}, {code=H314, sequence=4, description=Causes severe skin burns and eye damage.}, {code=H318, sequence=5, description=Causes serious eye damage.}, {code=H331, sequence=6, description=Toxic if inhaled.}, {code=H335, sequence=7, description=May cause respiratory irritation.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=54, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=56, description=Immediately call a POISON CENTER/doctor/.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Triethylamine}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}, {code=GHS06, sequence=3, description=SKULL & CROSSBONES}]
Product Identifier
- Triethylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H302- Harmful if swallowed.
- H311- Toxic in contact with skin.
- H314- Causes severe skin burns and eye damage.
- H318- Causes serious eye damage.
- H331- Toxic if inhaled.
- H335- May cause respiratory irritation.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
Supplemental Information
- MIXTURE LIST- Contains : Triethylamine