Use promo code "24156" to get your promotional price.
Sartorius Vivaspin™ 20 Centrifugal Concentrator
Offers increased volume flexibility and performance. VivaspinTM 20 Centrifugal Concentrator handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Features twin vertical membranes for unparalleled filtration speed and >100-fold concentration.
142.99 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Code:
53
Additional Details:
Weight:
0.38700kg
Specification
Disposable | Single-use |
---|---|
Filter Type | Ultrafiltration |
For Use With (Equipment) | Centrifuge |
Material (Membrane) | PES |
MWCO | 10 kDa |
Sample Volume | 20 mL |
Sterility | Non-sterile |
Quantity | 12/Pk. |
Type | Centrifugal Concentrator |
Volume (Metric) | 14 mL |
Length (Metric) | 116 mm |