Use promo code "24112" to get your promotional price.
Invitrogen™ PureLink™ Quick Plasmid Miniprep Kit
Use promo code "24112" to get your promotional price.
Specification
Concentration | 20 mg/mL |
---|---|
Column Type | Spin Column |
Content And Storage |
• Resuspension buffer • RNase A (20 mg/mL) • Lysis buffer • Precipitation buffer • Wash buffers • TE buffer • Spin columns in wash tubes • Elution tubes |
Format | Spin Column |
Isolation Technology | Silica Spin Column |
Scale | Mini |
Sample Type | Bacterial Culture |
Final Product Type | Plasmid DNA |
For Use With (Application) | Next-Generation Sequencing, Cloning, Sequencing, Nucleic Acid Labeling, PCR, Transformation |
High-throughput Compatibility | Not High-throughput Compatible (Manual) |
No. of Reactions | 250 Preps |
Prep Scale | < 100 μg (Small-Scale) Plasmid DNA |
Product Line | PureLink™ |
Product Type | Quick Plasmid MiniPrep Kit |
Quantity | 250 Preps |
Shipping Condition | Room Temperature |
Target | Plasmid DNA |
Test Time | 30 to 45 min. |
Technical documents
[PureLink Quick Plasmid Miniprep Kit]
[Warning]
[{code=EYE DAM. 2, sequence=1, description=Serious eye damage/eye irritation Category 2}, {code=SKIN IRRIT. 2, sequence=2, description=Skin corrosion/irritation Category 2}]
[{code=H315, sequence=1, description=Causes skin irritation.}, {code=H319, sequence=2, description=Causes serious eye irritation.}]
[{code=P264, sequence=51, description=Wash hands thoroughly after handling.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=53, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P332+P313, sequence=55, description=If skin irritation occurs: Get medical advice/attention.}, {code=P337+P313, sequence=56, description=If eye irritation persists: Get medical advice/attention.}, {code=P362+P364, sequence=57, description=Take off contaminated clothing and wash it before reuse.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Acetic acid}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}]
Product Identifier
- PureLink Quick Plasmid Miniprep Kit
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315- Causes skin irritation.
- H319- Causes serious eye irritation.
Precautionary Statement
- P264- Wash hands thoroughly after handling.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313- If skin irritation occurs: Get medical advice/attention.
- P337+P313- If eye irritation persists: Get medical advice/attention.
- P362+P364- Take off contaminated clothing and wash it before reuse.
Supplemental Information
- MIXTURE LIST- Contains : Acetic acid