730.40 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Use promo code "24074" to get your promotional price.
Content And Storage | Contains one vial (1.5 ml) Lipofectamine™ 2000 Reagent. Store at 2–8°C. Do not freeze. |
---|---|
Cell Type | Established Cell Lines,Stem Cells,Primary Cells,Hard-to-Transfect Cells |
Cell Type | Established Cell Lines,Stem Cells,Primary Cells,Hard-to-Transfect Cells |
Cell Type | Established Cell Lines,Stem Cells,Primary Cells,Hard-to-Transfect Cells |
Cell Type | Established Cell Lines,Stem Cells,Primary Cells,Hard-to-Transfect Cells |
Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
Sample Type | Plasmid DNA,Synthetic siRNA,RNAi Plasmids (shRNA, miR) |
Sample Type | Plasmid DNA,Synthetic siRNA,RNAi Plasmids (shRNA, miR) |
Sample Type | Plasmid DNA,Synthetic siRNA,RNAi Plasmids (shRNA, miR) |
Transfection Technique | Lipid-based Transfection |
For Use With (Application) | Transfection |
Green Features | Sustainable packaging |
High-throughput Compatibility | High-throughput Compatible |
Product Line | Lipofectamine™ |
Product Type | Transfection Reagent |
Quantity | 1.5 mL |
Serum Compatible | Yes |
Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |