Use promo code "25339" to get your promotional price.
Thermo Scientific Acros Di-tert-butyl dicarbonate, 99%
Use promo code "25339" to get your promotional price.
Quantity | 25 g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
CAS | 24424-99-5 |
---|---|
Molecular Formula | C10H18O5 |
Molecular Weight (g/mol) | 218.25 |
MDL Number | MFCD00008805 |
InChI Key | DYHSDKLCOJIUFX-UHFFFAOYSA-N |
Synonym | di-tert-butyl dicarbonate,boc anhydride,boc-anhydride,di-t-butyl dicarbonate,di-tert-butyl pyrocarbonate,bis tert-butoxycarbonyl oxide,di-tert-butyldicarbonate,boc2o,di-t-butyl pyrocarbonate,di tert-butyl carbonate |
PubChem CID | 90495 |
ChEBI | CHEBI:48500 |
IUPAC Name | tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate |
SMILES | CC(C)(C)OC(=O)OC(=O)OC(C)(C)C |
Specification
Chemical Name or Material | Di-tert-butyl dicarbonate |
---|---|
Name Note | 99% |
CAS | 24424-99-5 |
Melting Point | 22.0°C to 24.0°C |
Color | White |
Density | 1.0200g/mL |
Boiling Point | 56.0°C to 57.0°C (0.5 mmHg) |
Flash Point | 37°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Molecular Formula | C10H18O5 |
Linear Formula | O[CO2C(CH3)3]2 |
Refractive Index | 1.4075 to 1.4095 |
MDL Number | MFCD00008805 |
Quantity | 25 g |
Fieser | 04,128; 07,91; 08,145; 10,122; 12,159; 13,94; 15,113 |
Merck Index | 15, 3038 |
Specific Gravity | 1.02 |
Synonym | di-tert-butyl dicarbonate,boc anhydride,boc-anhydride,di-t-butyl dicarbonate,di-tert-butyl pyrocarbonate,bis tert-butoxycarbonyl oxide,di-tert-butyldicarbonate,boc2o,di-t-butyl pyrocarbonate,di tert-butyl carbonate |
Synonym | di-tert-butyl dicarbonate,boc anhydride,boc-anhydride,di-t-butyl dicarbonate,di-tert-butyl pyrocarbonate,bis tert-butoxycarbonyl oxide,di-tert-butyldicarbonate,boc2o,di-t-butyl pyrocarbonate,di tert-butyl carbonate |
Synonym | di-tert-butyl dicarbonate,boc anhydride,boc-anhydride,di-t-butyl dicarbonate,di-tert-butyl pyrocarbonate,bis tert-butoxycarbonyl oxide,di-tert-butyldicarbonate,boc2o,di-t-butyl pyrocarbonate,di tert-butyl carbonate |
Synonym | di-tert-butyl dicarbonate,boc anhydride,boc-anhydride,di-t-butyl dicarbonate,di-tert-butyl pyrocarbonate,bis tert-butoxycarbonyl oxide,di-tert-butyldicarbonate,boc2o,di-t-butyl pyrocarbonate,di tert-butyl carbonate |
Synonym | di-tert-butyl dicarbonate,boc anhydride,boc-anhydride,di-t-butyl dicarbonate,di-tert-butyl pyrocarbonate,bis tert-butoxycarbonyl oxide,di-tert-butyldicarbonate,boc2o,di-t-butyl pyrocarbonate,di tert-butyl carbonate |
Synonym | di-tert-butyl dicarbonate,boc anhydride,boc-anhydride,di-t-butyl dicarbonate,di-tert-butyl pyrocarbonate,bis tert-butoxycarbonyl oxide,di-tert-butyldicarbonate,boc2o,di-t-butyl pyrocarbonate,di tert-butyl carbonate |
Synonym | di-tert-butyl dicarbonate,boc anhydride,boc-anhydride,di-t-butyl dicarbonate,di-tert-butyl pyrocarbonate,bis tert-butoxycarbonyl oxide,di-tert-butyldicarbonate,boc2o,di-t-butyl pyrocarbonate,di tert-butyl carbonate |
Synonym | di-tert-butyl dicarbonate,boc anhydride,boc-anhydride,di-t-butyl dicarbonate,di-tert-butyl pyrocarbonate,bis tert-butoxycarbonyl oxide,di-tert-butyldicarbonate,boc2o,di-t-butyl pyrocarbonate,di tert-butyl carbonate |
Synonym | di-tert-butyl dicarbonate,boc anhydride,boc-anhydride,di-t-butyl dicarbonate,di-tert-butyl pyrocarbonate,bis tert-butoxycarbonyl oxide,di-tert-butyldicarbonate,boc2o,di-t-butyl pyrocarbonate,di tert-butyl carbonate |
Synonym | di-tert-butyl dicarbonate,boc anhydride,boc-anhydride,di-t-butyl dicarbonate,di-tert-butyl pyrocarbonate,bis tert-butoxycarbonyl oxide,di-tert-butyldicarbonate,boc2o,di-t-butyl pyrocarbonate,di tert-butyl carbonate |
InChI Key | DYHSDKLCOJIUFX-UHFFFAOYSA-N |
SMILES | CC(C)(C)OC(=O)OC(=O)OC(C)(C)C |
IUPAC Name | tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate |
Molecular Weight (g/mol) | 218.25 |
PubChem CID | 90495 |
ChEBI | CHEBI:48500 |
Formula Weight | 218.25 |
Percent Purity | 99% |
Physical Form | Low Melting Crystalline Solid |
Technical documents
[Di-tert-butyl dicarbonate]
[Danger]
[{code=ACUTE TOX. 1, sequence=1, description=Acute toxicity Category 1}, {code=EYE DAM. 1, sequence=2, description=Serious eye damage/eye irritation Category 1}, {code=FLAM. SOL. 2, sequence=3, description=Flammable solid Category 2}, {code=RESP. SENS. 1, sequence=4, description=Respiratory sensitiser Category 1}, {code=SKIN IRRIT. 2, sequence=5, description=Skin corrosion/irritation Category 2}, {code=STOT SE 3, sequence=6, description=Specific target organ toxicity Category 3}]
[{code=H317, sequence=1, description=May cause an allergic skin reaction.}, {code=H318, sequence=2, description=Causes serious eye damage.}, {code=H330, sequence=3, description=Fatal if inhaled.}, {code=H315, sequence=4, description=Causes skin irritation.}, {code=H335, sequence=5, description=May cause respiratory irritation.}, {code=H228, sequence=6, description=Flammable solid.}]
[{code=P210, sequence=50, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=52, description=IF ON SKIN: Wash with plenty of water/.}, {code=P304+P340, sequence=53, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=55, description=Immediately call a POISON CENTER/doctor/.}, {code=P332+P313, sequence=56, description=If skin irritation occurs: Get medical advice/attention.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}, {code=GHS06, sequence=3, description=SKULL & CROSSBONES}]
Product Identifier
- Di-tert-butyl dicarbonate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317- May cause an allergic skin reaction.
- H318- Causes serious eye damage.
- H330- Fatal if inhaled.
- H315- Causes skin irritation.
- H335- May cause respiratory irritation.
- H228- Flammable solid.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
- P332+P313- If skin irritation occurs: Get medical advice/attention.