Code:
50
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column
Additional Details:
Weight:
0.01000kg
Specification
Diameter (Metric) | 0.4 mm |
---|---|
Type | GC Capillary Column |
Diameter (Metric) Inner | 0.25 mm |
Stationary Phase | Permabond™ FFAP |
USP Type | G25, G35 |
For Use With (Application) | Gas Chromatography (GC),Free Carboxylic Acids |
For Use With (Application) | Gas Chromatography (GC),Free Carboxylic Acids |
Temperature Range | 220°C to 240°C |
Film Thickness | 0.1 μm |
Length (Metric) | 25 m |
Packing Material | Permabond™ FFAP |
Quantity | 1 Each |