323.33 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Max. Temperature | 240/250°C |
---|---|
Diameter (Metric) | 0.25 mm |
Diameter (Metric) Inner | 0.25 mm |
Product Line | TRACE |
Stationary Phase | Trace™ TR-FFAP |
USP Type | G25, G35 |
For Use With (Application) | Acidic Compound Analysis,GC for Acidic Compound Analysis |
For Use With (Application) | Acidic Compound Analysis,GC for Acidic Compound Analysis |
Film Thickness | 0.25 μm |
Length (Metric) | 15 m |
Polarity | Polar |
Description | 15 m x 0.25 mm x 0.25 μm |