Fisher Chemical Karl Fischer Aqualine™ Electrolyte AG, for Karl Fischer Titration By Coulometry, Fisher Chemical™
Nonhalogenated anolyte solution for general use in conventional cells with a diaphragm
Overview
136.64 GBP valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Code: 24
Additional Details:
Additional Details: CAS Number: 108-75-8 Weight: 0.87000kg Transport: UN number : 1992 Chem class : 3 Pack group : II
Product Code. 10724893
Quantity Price
1 £ 207.0 / 500mL
In stock, ready to ship: 325
Add to basket
Description and Specification
Specification
Chemical Name or Material Aqualine™ Electrolyte AG
Color Colorless
Name Note Karl Fischer Reagent
Quantity 500 mL
CAS 108-75-8
Packaging Amber glass bottle
CAS 111-42-2
Physical Form Liquid
CAS 7446-09-5
CAS 7553-56-2
Use Test Pass Test
MDL Number 3097387
  • Anolyte solution for use in conventional cells with a diaphragm
  • Source of electrolyte required for anodic oxidation for coulometric analysis
  • Contains methanol as a solvent
Documentation Special Offers