274.36 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Array | 4 x 6 |
---|---|
Closure Included | With Lid |
Bottom Shape | Flat |
Capacity (Metric) | 3.4 mL |
Color | Clear |
Culture Area | 1.9 cm2 |
Material | Polystyrene |
Sterility | Sterile |
Surface Treatment | Tissue Culture |
Type | Cell Culture Microplate |
Volume (Metric) Working | 0.5 mL |
Well Count | 24 |
No. per Case | 100 (Individually wrapped) |
No. per Pack | Individually Wrapped |
Packaging | Individually Wrapped |
Quantity | 100/Cs. |
Diameter (Metric) | 1.6 cm |