Use promo code "25339" to get your promotional price.
Fisher Chemical Butan-1-ol, Certified AR for Analysis, Fisher Chemical™
Use promo code "25339" to get your promotional price.
Quantity | 25 L |
---|---|
Packaging | Metal Can |
Chemical Identifiers
CAS | 71-36-3 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | 2964 |
InChI Key | LRHPLDYGYMQRHN-UHFFFAOYSA-N |
IUPAC Name | butan-1-ol |
SMILES | CCCCO |
Specification
Melting Point | -89°C |
---|---|
CAS | 71-36-3 |
Color | Colorless |
Boiling Point | 117.6°C |
Quantity | 25 L |
Packaging | Metal Can |
Physical Form | Liquid |
Molecular Formula | C4H10O |
MDL Number | 2964 |
Vapor Pressure | 6.7mbar at 20°C |
Viscosity | 2.95 mPaS ( at 20°C) |
InChI Key | LRHPLDYGYMQRHN-UHFFFAOYSA-N |
SMILES | CCCCO |
IUPAC Name | butan-1-ol |
Molecular Weight (g/mol) | 74.12 |
Formula Weight | 74.12g/mol |
Technical documents
[Butan-1-ol]
[Danger]
[{code=FLAM. LIQ. 3, sequence=1, description=Flammable liquid Category 3}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=SKIN IRRIT. 2, sequence=3, description=Skin corrosion/irritation Category 2}, {code=EYE DAM. 1, sequence=4, description=Serious eye damage/eye irritation Category 1}, {code=STOT SE 3, sequence=5, description=Specific target organ toxicity Category 3}]
[{code=H226, sequence=1, description=Flammable liquid and vapour.}, {code=H302, sequence=2, description=Harmful if swallowed.}, {code=H315, sequence=3, description=Causes skin irritation.}, {code=H318, sequence=4, description=Causes serious eye damage.}, {code=H335, sequence=5, description=May cause respiratory irritation.}, {code=H336, sequence=6, description=May cause drowsiness or dizziness.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P303+P361+P353, sequence=53, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=54, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=56, description=Immediately call a POISON CENTER/doctor/.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}, {code=GHS07, sequence=3, description=EXCLAMATION MARK}]
Product Identifier
- Butan-1-ol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226- Flammable liquid and vapour.
- H302- Harmful if swallowed.
- H315- Causes skin irritation.
- H318- Causes serious eye damage.
- H335- May cause respiratory irritation.
- H336- May cause drowsiness or dizziness.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.