Macherey-Nagel™ MN 715 Grade Filter Papers, Folded
Particularly suited for fat analysis. Macherey-Nagel™ MN 715 Grade Filter Papers, Folded are smooth qualitative papers made of carefully selected raw materials to ensure fat-free paper with a low ether-soluble residue.
Additional Details:
Weight:
0.10000kg
Specification
Basis Weight | 70 g/m2 |
---|---|
Description | Smooth, fat-free |
Format | Folded |
Material | Cellulose |
Filtration Speed | 22sec. |
For Use With (Application) | Analysis of fats |
Grade | MN 715 |
Quantity | 100 Pack |
Thickness (Metric) | 0.16 mm |
Type | Filter Paper |
Diameter (Metric) | 125 mm |