151.72 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Certifications/Compliance | Certified non-pyrogenic |
---|---|
Closure Material | HDPE |
Closure Color | Orange |
Color | Natural |
Capacity (Metric) | 250 mL |
Disposable | Single-use |
Material | Polypropylene |
No. per Case | 102 |
No. per Pack | 6 |
Packaging | No rack |
Sterility | Sterile |
Shape | Conical |
Length (Metric) | 159 mm |
Capacity (English) | 8.45 oz. |
Bottom Shape | Conical |
Closure Type | Plug |
Writing Area | No |
Diameter (Metric) | 60 mm |
Graduated | Yes |
Product Type | Centrifuge Tube |
Max. RCF | 6000 x g |