72.89 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Use promo code "24123" to get your promotional price.
Certifications/Compliance | ISO 1773, DIN 12353 |
---|---|
Capacity (Metric) | 5 L |
Material | Borosilicate Glass,Borosilicate Glass |
Material | Borosilicate Glass,Borosilicate Glass |
Graduations | Yes |
Height (Metric) | 365 mm |
Shape | Conical |
Writing Surface | Yes |
Graduated | Yes |
Diameter (Metric) Outer | 220 mm |
Closure Size | 45 |