Use promo code "25339" to get your promotional price.
Thermo Scientific Acros tert-Butyl methyl ether, 99%, pure
Use promo code "25339" to get your promotional price.
Quantity | 250 mL |
---|---|
Packaging | Glass Bottle |
Chemical Identifiers
CAS | 1634-04-4 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
MDL Number | MFCD00008812 |
InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
PubChem CID | 15413 |
ChEBI | CHEBI:27642 |
IUPAC Name | 2-methoxy-2-methylpropane |
SMILES | CC(C)(C)OC |
Specification
Chemical Name or Material | tert-Butyl methyl ether |
---|---|
Melting Point | -110.0°C |
Name Note | 99% |
CAS | 1634-04-4 |
Color | White to Yellow |
Density | 0.7404g/mL |
Boiling Point | 54.0°C to 56.0°C |
Quantity | 250 mL |
Flash Point | −28°C |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Physical Form | Powder or Needles |
Refractive Index | 1.3675 to 1.3695 |
Assay Percent Range | 99% |
Specific Gravity | 0.7404 |
Molecular Formula | C5H12O |
Linear Formula | (CH3)3COCH3 |
MDL Number | MFCD00008812 |
Viscosity | 0.36 mPa.s (20°C) |
Beilstein | 01, 381 |
Water | 0.01% Max. (Coulometric) |
Fieser | 04,333 |
Merck Index | 15, 6106 |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Synonym | tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane |
Solubility Information | Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g |
InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
SMILES | CC(C)(C)OC |
IUPAC Name | 2-methoxy-2-methylpropane |
Molecular Weight (g/mol) | 88.15 |
PubChem CID | 15413 |
ChEBI | CHEBI:27642 |
Formula Weight | 88.15 |
Percent Purity | 99% |
Grade | Pure |
Technical documents
[tert-Butyl methyl ether]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}, {code=SKIN IRRIT. 2, sequence=2, description=Skin corrosion/irritation Category 2}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H315, sequence=2, description=Causes skin irritation.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P240, sequence=52, description=Ground/bond container and receiving equipment.}, {code=P302+P352, sequence=53, description=IF ON SKIN: Wash with plenty of water/soap.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- tert-Butyl methyl ether
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H315- Causes skin irritation.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240- Ground/bond container and receiving equipment.
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.