Kautex™ Red Polypropylene Screw Closures
Seal a container's ingredients with these Kautex™ Red Polypropylene Screw Closures when you package, send samples, transport and store their contents.
Additional Details:
Weight:
0.02790kg
Specification
Color | Red |
---|---|
Includes | Tamper Evident Ring and Polyethylene Foam Insert |
Material | Polypropylene |
Liner Material | Polyethylene Foam Insert |
Product Type | Screw Cap |
For Use With (Equipment) | 1500/2500/4000 mL Bottle |
Diameter (Metric) | 80 mm |