61.61 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Chemical Name or Material | Sodium hydroxide |
---|---|
Color | Colorless |
CAS | 1310-73-2 |
CAS | 7732-18-5 |
Molecular Formula | HNaO |
Linear Formula | NaOH |
Concentration | 0.998 to 1.002N (20°C) |
Fieser | 01,1083; 05,616; 07,336; 08,460 |
Density | 1.0400g/mL |
Merck Index | 15, 8761 |
Synonym | sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic |
Synonym | sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic |
Synonym | sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic |
Synonym | sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic |
Synonym | sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic |
Synonym | sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic |
Synonym | sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic |
Synonym | sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic |
Synonym | sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic |
Synonym | sodium hydroxide,caustic soda,sodium hydrate,white caustic,soda lye,aetznatron,ascarite,sodium hydroxide na oh,sodium hydroxide solution,soda, caustic |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol and glycerol |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Quantity | 5 L |
SMILES | [OH-].[Na+] |
IUPAC Name | sodium;hydroxide |
Molecular Weight (g/mol) | 39.997 |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
Formula Weight | 40 |
Packaging | Plastic drum |
Specific Gravity | 1.04 |
Physical Form | Liquid |