72.00 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Concentration | 6X |
---|---|
Gel Type | Agarose |
Label or Dye | Xylene Cyanol FF, Orange G |
Product Type | DNA Ladder |
Ready to Load | Yes |
Size Range | 100 to 6000 bp |
Gel Compatibility | Agarose Gels |
Green Features | Sustainable packaging |
Product Line | O'GeneRuler, O’RangeRuler |
Quantity | 25 μg |