Chemical Name or Material |
Palladium (II) chloride
|
Melting Point |
675°C (decomposition)
|
CAS |
7647-10-1
|
Color |
Red to Brown
|
Physical Form |
Powder
|
Quantity |
25 g
|
Assay Percent Notes |
(metals basis)
|
Molecular Formula |
Cl2Pd
|
MDL Number |
MFCD00003558
|
Merck Index |
14,6990
|
Synonym |
palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440
|
Synonym |
palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440
|
Synonym |
palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440
|
Synonym |
palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440
|
Synonym |
palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440
|
Synonym |
palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440
|
Synonym |
palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440
|
Synonym |
palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440
|
Solubility Information |
Soluble in cold water,organic solvents and hydrochloric acid.
|
InChI Key |
PIBWKRNGBLPSSY-UHFFFAOYSA-L
|
SMILES |
[Cl-].[Cl-].[Pd++]
|
IUPAC Name |
palladium(2+) dichloride
|
Molecular Weight (g/mol) |
177.32
|
PubChem CID |
24290
|
ChEBI |
CHEBI:53434
|
Formula Weight |
177.31
|
Percent Purity |
99.999%
|
Grade |
Premion™
|
Concentration or Composition (by Analyte or Components) |
Pd 59.5% min
|
Odor |
Odorless
|
Density |
4.0 g/mL
|