Chemical Name or Material |
Ethylenediaminetetraacetic acid
|
CAS |
60-00-4
|
Melting Point |
∼245°C (decomposition)
|
Color |
White
|
Molecular Formula |
C10H16N2O8
|
Linear Formula |
(HO2CCH2)2NCH2CH2N(CH2CO2H)2
|
MDL Number |
MFCD00003541
|
Quantity |
2.5 kg
|
Beilstein |
1716295
|
Merck Index |
14,3517
|
Synonym |
edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol
|
Synonym |
edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol
|
Synonym |
edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol
|
Synonym |
edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol
|
Synonym |
edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol
|
Synonym |
edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol
|
Synonym |
edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol
|
Synonym |
edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol
|
Synonym |
edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol
|
Synonym |
edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol
|
Solubility Information |
Soluble in sodium hydroxide,sodium carbonate,ammonia solution,acid and common organic solvents. Slightly soluble in water.
|
InChI Key |
KCXVZYZYPLLWCC-UHFFFAOYSA-N
|
SMILES |
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
|
IUPAC Name |
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
|
Molecular Weight (g/mol) |
292.24
|
PubChem CID |
6049
|
ChEBI |
CHEBI:42191
|
Formula Weight |
292.24
|
Percent Purity |
≥99.4%
|
Grade |
ACS
|
Physical Form |
Powder
|