68.64 GBP valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Use promo code "24140" to get your promotional price.
Quantity | 1000 g |
---|
CAS | 308080-99-1 |
---|---|
Molecular Formula | AlNaO6Si2 |
Molecular Weight (g/mol) | 202.135 |
MDL Number | MFCD00131613 |
InChI Key | URGAHOPLAPQHLN-UHFFFAOYSA-N |
PubChem CID | 19758701 |
IUPAC Name | aluminum;sodium;dioxido(oxo)silane |
SMILES | [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] |
Chemical Name or Material | Molecular sieves, 3A |
---|---|
Name Note | 3 to 5mm (0.12 to 0.20 in) beads |
Sensitivity | Hygroscopic |
Quantity | 1000 g |
CAS | 308080-99-1 |
Molecular Formula | AlNaO6Si2 |
MDL Number | MFCD00131613 |
Solubility Information | Insoluble in cold water,hot water,methanol,diethyl ether,n-octanol and acetone. |
InChI Key | URGAHOPLAPQHLN-UHFFFAOYSA-N |
SMILES | [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] |
IUPAC Name | aluminum;sodium;dioxido(oxo)silane |
Molecular Weight (g/mol) | 202.135 |
PubChem CID | 19758701 |