Use promo code "25339" to get your promotional price.
Thermo Scientific Alfa Aesar Acetic acid, glacial, 99+%, Thermo Scientific Chemicals
Use promo code "25339" to get your promotional price.
Quantity | 1 L |
---|
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Specification
Chemical Name or Material | Acetic acid |
---|---|
Name Note | glacial |
CAS | 64-19-7 |
Melting Point | 16.6°C |
Density | 1.049 |
Boiling Point | 118.1°C |
Flash Point | 40°C (104°F) |
Assay Percent Range | 99+% |
Molecular Formula | C2H4O2 |
Refractive Index | 1.3721 |
MDL Number | MFCD00036152 |
Quantity | 1 L |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Synonym | ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure |
Solubility Information | Miscible with water,ethanol,acetone,toluene and immiscible with carbon disulphide. |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
SMILES | CC(O)=O |
IUPAC Name | acetic acid |
Molecular Weight (g/mol) | 60.05 |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
Formula Weight | 60.05 |
Physical Form | Liquid |
Technical documents
[Acetic acid, glacial]
[Danger]
[{code=EYE DAM. 1, sequence=1, description=Serious eye damage/eye irritation Category 1}, {code=FLAM. LIQ. 3, sequence=2, description=Flammable liquid Category 3}, {code=SKIN CORR. 1A, sequence=3, description=Skin corrosion/irritation Category 1 A}]
[{code=H226, sequence=1, description=Flammable liquid and vapour.}, {code=H314, sequence=2, description=Causes severe skin burns and eye damage.}, {code=H318, sequence=3, description=Causes serious eye damage.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=54, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=56, description=Immediately call a POISON CENTER/doctor/.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : acetic acid}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}]
Product Identifier
- Acetic acid, glacial
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H226- Flammable liquid and vapour.
- H314- Causes severe skin burns and eye damage.
- H318- Causes serious eye damage.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
Supplemental Information
- MIXTURE LIST- Contains : acetic acid