Thermo Scientific Alfa Aesar Sodium cyanide, ACS, 95% min, Thermo Scientific Chemicals
Overview
26.60 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: CAS Number: 143-33-9 Weight: 0.10000kg Transport: UN number : 1689 Chem class : 6.1 Pack group : I
Quantity 100 g
Disclaimers: RUO – Research Use Only
Product Code. 11326449
Quantity Price
1 £ 26.73 / 100g
In stock, ready to ship: 2
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Chemical Identifiers
CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
Synonym sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
PubChem CID 8929
IUPAC Name sodium;cyanide
SMILES [Na+].[C-]#N
Specification
Boiling Point ca 1500°C
Chemical Name or Material Sodium cyanide
Melting Point 563°C
Name Note For analysis ACS
CAS 143-33-9
Color Gray
Physical Form Granules
Quantity 100 g
Assay Percent Range 95% min.
Molecular Formula CNNa
MDL Number MFCD00003523
Beilstein 3587243
Synonym sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
Synonym sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
Synonym sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
Synonym sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
Synonym sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
Synonym sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
Synonym sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
Synonym sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
Synonym sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
Synonym sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h
Solubility Information Soluble in water,ammonia,methanol and ethanol. Slightly soluble in dimethylformamide. Insoluble in dimethylsulphoxide.
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
SMILES [Na+].[C-]#N
IUPAC Name sodium;cyanide
Molecular Weight (g/mol) 49.01
PubChem CID 8929
Formula Weight 49.01
Grade ACS
Odor Weak, characteristic
Sensitivity Hygroscopic
Appearance White powder
Density 1.61 g/mL