Code:
69
Cytiva Whatman™ Litmus Blue Test Paper
Impregnated with a mixture of pH indicators, acid-alkali litmus papers assume a color specific to sample pH upon contact with a sample. Whatman™ Litmus Blue Test Paper provides rapid determination of pH values and has a variety of applications in laboratories and industry.
Additional Details:
Weight:
0.07200kg
Specification
Color Indicator | Blue |
---|---|
pH Range | 5 to 8 |
Format | Strip |
Quantity | 100 Pack |
Type | Test Paper |