Chemical Name or Material |
Potassium cyanate
|
Melting Point |
700°C to 900°C (decomposition)
|
CAS |
590-28-3
|
Color |
White
|
Physical Form |
Crystalline
|
Quantity |
2 kg
|
Molecular Formula |
CKNO
|
MDL Number |
MFCD00011411
|
Merck Index |
14,7625
|
Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
Solubility Information |
Soluble in water; Slightly soluble in alcohol
|
InChI Key |
GKKCIDNWFBPDBW-UHFFFAOYSA-M
|
SMILES |
[K+].[O-]C#N
|
IUPAC Name |
potassium cyanate
|
Molecular Weight (g/mol) |
81.12
|
PubChem CID |
11378442
|
ChEBI |
CHEBI:38904
|
Formula Weight |
81.12
|
Percent Purity |
97%
|
Odor |
Odorless
|
Density |
2.056 g/mL
|