Code:
A0
Axygen™ 1.5 mL Conical Screw Cap Tubes
With chemical-resistant O-ring
Additional Details:
Weight:
0.90900kg
Specification
Cap Style | Screw Cap |
---|---|
Certifications/Compliance | RNase, DNase, pyrogen-free |
Closure Color | Clear |
Closure Material | Polypropylene |
Capacity (Metric) | 1.5 mL |
Color | Clear |
Material | Polypropylene |
Format | Non-looped |
Includes | O-ring |
No. per Pack | 500/Pk. |
Sterility | Sterile |
Graduated | No |
Closure Type | Screw Cap |
Bottom Shape | Conical |
Max. RCF | 20,000 x g |