Use promo code "25339" to get your promotional price.
Thermo Scientific Alfa Aesar Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals
Use promo code "25339" to get your promotional price.
Quantity | 100 g |
---|
Chemical Identifiers
CAS | 60-00-4 |
---|---|
Molecular Formula | C10H16N2O8 |
Molecular Weight (g/mol) | 292.24 |
MDL Number | MFCD00003541 |
InChI Key | KCXVZYZYPLLWCC-UHFFFAOYSA-N |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
PubChem CID | 6049 |
ChEBI | CHEBI:42191 |
IUPAC Name | 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid |
SMILES | OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Specification
Chemical Name or Material | Ethylenediaminetetraacetic acid |
---|---|
CAS | 60-00-4 |
Melting Point | ∼245°C (decomposition) |
Color | White |
Molecular Formula | C10H16N2O8 |
Linear Formula | (HO2CCH2)2NCH2CH2N(CH2CO2H)2 |
MDL Number | MFCD00003541 |
Quantity | 100 g |
Beilstein | 1716295 |
Merck Index | 14,3517 |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Synonym | edta,edetic acid,ethylenediaminetetraacetic acid,edathamil,versene,endrate,havidote,titriplex,edta acid,sequestrol |
Solubility Information | Soluble in sodium hydroxide,sodium carbonate,ammonia solution,acid and common organic solvents. Slightly soluble in water. |
InChI Key | KCXVZYZYPLLWCC-UHFFFAOYSA-N |
SMILES | OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
IUPAC Name | 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid |
Molecular Weight (g/mol) | 292.24 |
PubChem CID | 6049 |
ChEBI | CHEBI:42191 |
Formula Weight | 292.24 |
Percent Purity | ≥99.4% |
Grade | ACS |
Physical Form | Powder |
Technical documents
[Ethylenediaminetetraacetic acid, ACS]
[Warning]
[{code=H319, sequence=1, description=Causes serious eye irritation.}, {code=H332, sequence=2, description=Harmful if inhaled.}, {code=H373, sequence=3, description=May cause damage to organs.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P304+P340, sequence=52, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=53, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P312, sequence=54, description=Call a POISON CENTER/doctor/if you feel unwell.}, {code=P337+P313, sequence=55, description=If eye irritation persists: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Ethylenediaminetetraacetic acid}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}, {code=GHS08, sequence=2, description=HEALTH HAZARD}]
Product Identifier
- Ethylenediaminetetraacetic acid, ACS
Signal Word
- Warning
Hazard Statement
- H319- Causes serious eye irritation.
- H332- Harmful if inhaled.
- H373- May cause damage to organs.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312- Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313- If eye irritation persists: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contains : Ethylenediaminetetraacetic acid